Learn More
AMMECR1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 246871AP
Catalog No. 50-173-4488
Add to cart
Description
Description
The exact function of this gene is not known, however, submicroscopic deletion of the X chromosome including this gene, COL4A5, and FACL4 genes, result in a contiguous gene deletion syndrome, the AMME complex (Alport syndrome, mental retardation, midface hypoplasia, and elliptocytosis). Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
AMMECR1
0.45 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9JHT5, Q9Y4X0
AMMECR1, AMMERC1
Rabbit
IgG
150 μL
-20°C
Primary
56068, 9949
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
AMMECR1
Liquid
Ammecr1
AMMECR1 Fusion Protein Ag16977
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse