Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonia, 0.5M solution in 1,4-dioxane, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
1L
Packaging:
AcroSeal™ Glass bottle
7664-41-7,123-91-1
H3N
17.031
MFCD00011418
QGZKDVFQNNGYKY-UHFFFAOYSA-N
ammonia,ammonia gas,spirit of hartshorn,nitro-sil,ammoniakgas,ammonia anhydrous,anhydrous ammonia,ammonia, anhydrous,ammoniak,am-fol
222
CHEBI:16134
azane
N
7664-41-7,123-91-1
H3N
17.031
MFCD00011418
QGZKDVFQNNGYKY-UHFFFAOYSA-N
ammonia,ammonia gas,spirit of hartshorn,nitro-sil,ammoniakgas,ammonia anhydrous,anhydrous ammonia,ammonia, anhydrous,ammoniak,am-fol
222
CHEBI:16134
azane
N
Linear Formula NH3
Concentration 0.4 to 0.6M
Fieser 05,15
Merck Index 15, 488
Density 1.0230g/mL
Solubility Information Solubility in water: soluble.
Formula Weight 17.03
Physical Form Liquid
Packaging AcroSeal™ Glass bottle
Solubility Solubility in water: soluble.
Specific Gravity 1.023
Water 1% max. (K.F.)
Color Colorless
Quantity 100mL
Flash Point 11°C
Chemical Name or Material Ammonia, 0.5M solution in 1, 4-dioxane
Show More Show Less

GHS H Statement
Highly flammable liquid and vapor. Causes serious eye irritation. May cause respiratory irritation. Suspected of causing cancer. Repeated exposure may cause skin dryness or cracking.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Do not breathe dust/fume/gas/mist/vapors/spray. Wash face, hands and any exposed skin thoroughly after handling. Use personal protective equipment as required. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

Danger