Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonia-Ammonium Chloride Buffer TS, Reagents

Catalog No. 01336047
Click to view available options
Quantity:
500 mL
1 L
20 L

Catalog No. 01-336-047

Supplier: Reagents Holdings Llc CA01430020F

Only null left
Add to Cart
Add to Cart

Ammonia-Ammonium Chloride Buffer TS, Meets USP specifications, use for EDTA titrations. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Ammonia-Ammonium Chloride Buffer TS
CAS 7732-18-5, 1336-21-6, 1336-21-6
Boiling Point 36°C
Color Colorless
For Use With (Application) Titration
pH 10
Physical Form Liquid
Quantity 20 L
Solution Type Buffer
Packaging Cubitainer
Specific Gravity 0.9
Sterility Non-sterile
Grade USP
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.