Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonia buffer solution, Ammonium chloride / ammonia, pH 10, for complexometry, Solstice

Catalog No. p-7122060 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
6 x 1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-002-96 1 L
60-046-284 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-002-96 Supplier Solstice Supplier No. 335821L
Add to Cart
Add to Cart

Ammonium chloride / ammonia, pH 10, for complexometry

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ammonia buffer solution
CAS 12125-02-9
Quantity 1 L
Molecular Formula ClH4N
Packaging Poly bottle
MDL Number MFCD00011420
UN Number NONH for all modes of transport
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
SMILES N.Cl
IUPAC Name azanium;chloride
Molecular Weight (g/mol) 53.49
PubChem CID 25517
ChEBI CHEBI:31206
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.