Learn More
Ammonia, ca. 7N solution in methanol, Thermo Scientific Chemicals
Ammonia, H3N, CAS Number-7664-41-7, 67-56-1, am-fol, ammonia, ammonia anhydrous, ammonia, anhydrous, ammonia gas, ammoniak, ammoniakgas, anhydrous ammonia, nitro-sil, spirit of hartshorn, 2.5L, CHEBI:16134, Colorless to Yellow, 6.0 to 7.5N, 0.7700g/mL, 05,15, 14 deg.C, 17.03, GHS Signal Word: Danger
Supplier: Thermo Scientific Chemicals 133710250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ammonia | |
ca. 7N Solution in Methanol | |
H3N | |
6.0 to 7.5N | |
05,15 | |
0.7700g/mL | |
25 mL | |
N | |
17.03 | |
14°C | |
17.03 | |
0.77 |
Colorless to Yellow | |
7664-41-7, 67-56-1 | |
NH3 | |
MFCD00011418 | |
15, 488 | |
ammonia, ammonia gas, spirit of hartshorn, nitro-sil, ammoniakgas, ammonia anhydrous, anhydrous ammonia, ammonia, anhydrous, ammoniak, am-fol | |
QGZKDVFQNNGYKY-UHFFFAOYSA-N | |
azane | |
222 | |
CHEBI:16134 | |
Glass Bottle | |
Liquid |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Causes damage to organs.
Fatal if inhaled.
Highly flammable liquid and vapor.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF exposed: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance
GHS Signal Word: Danger
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
RUO – Research Use Only