Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonia, ca. 7N solution in methanol, ACROS Organics™

Click to view available options
Quantity:
25mL
1L
2.5L
25L
Packaging:
Glass Bottle
Plastic drum
7664-41-7,67-56-1
H3N
17.031
MFCD00011418
QGZKDVFQNNGYKY-UHFFFAOYSA-N
ammonia,ammonia gas,spirit of hartshorn,nitro-sil,ammoniakgas,ammonia anhydrous,anhydrous ammonia,ammonia, anhydrous,ammoniak,am-fol
222
CHEBI:16134
azane
N
7664-41-7,67-56-1
H3N
17.031
MFCD00011418
QGZKDVFQNNGYKY-UHFFFAOYSA-N
ammonia,ammonia gas,spirit of hartshorn,nitro-sil,ammoniakgas,ammonia anhydrous,anhydrous ammonia,ammonia, anhydrous,ammoniak,am-fol
222
CHEBI:16134
azane
N
Color Colorless to Yellow
Flash Point 14°C
Quantity 25L
Concentration 6.0 to 7.5N
Linear Formula NH3
Density 0.7700g/mL
Fieser 05,15
Merck Index 15, 488
Formula Weight 17.03
Physical Form Liquid
Packaging Plastic drum
Specific Gravity 0.77
Water 0.5% max.
Chemical Name or Material Ammonia, ca. 7N Solution in Methanol
Show More Show Less

GHS H Statement:
Toxic if swallowed.
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Causes damage to organs.
Fatal if inhaled.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF exposed: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger