Learn More
Brady™ Ammonia (IIAR) Pipe Markers, Legend: HSS/VAP/LOW/Ammonia, B-946 Self-Sticking Vinyl
For use in labeling ammonia piping and its contents. Brady Ammonia (IIAR) Pipe Markers help create a safer, more efficient work environment.
$6.10 - $6.95
Specifications
Product Type Ammonia (IIAR) Pipe Marker
Color Black, Blue, Green, Yellow
Format Die-cut on cards
Adhesive Type Acrylic, Cold Temperature, Permanent
Material Pressure Sensitive Vinyl (B-946)
View More Specs
Products 3
Catalog Number Mfr. No. Description Diameter (Metric) Height (Metric) Character Price Quantity & Availability  
Catalog Number Mfr. No. Description Diameter (Metric) Height (Metric) Character Price Quantity & Availability  
19-110-552 View Documents Brady™
90408
Style I 76.2 x 127 mm 50.8 mm Each for $6.10
Add to cart
19-110-572 View Documents Brady™
90428
Style 4 25.4 x 63.5 mm 19.05 mm Each for $6.10
Add to cart
19-110-592 View Documents Brady™
90448
Style IHV 203.2 mm 88.9 mm Each for $6.95
Add to cart
Description
Description
  • Meets IIAR Standard and is recommended for use in labeling ammonia piping and its contents
  • Made of B-946 Self sticking pressure sensitive vinyl
  • Available in three different style codes
Specifications
Specifications
Ammonia (IIAR) Pipe Marker
Die-cut on cards
Pressure Sensitive Vinyl (B-946)
Pipe and Valve marking
Black, Blue, Green, Yellow
Acrylic, Cold Temperature, Permanent
IIAR Abbreviation: HSS; Legend: Ammonia; Physical State: VAP; Pressure Level: LOW