Learn More
Brady™ Ammonia (IIAR) Pipe Markers, Legend: LIQ/Ammonia/HIGH
For use in labeling ammonia piping and its contents. Brady Ammonia (IIAR) Pipe Markers help create a safer, more efficient work environment.
Supplier:  Brady™ 59939
 View more versions of this product
Catalog No. 19-110-528
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative.
Description
Description
  • Meets IIAR Standard and is recommended for use in labeling ammonia piping and its contents
  • Made of B-689 high performance wrap-around PVF over-laminated polyester and B-681/B-883 polyester over-laminate on fiberglass plastic carrier
Specifications
Specifications
Style HPHV
Polyester Over-laminate (B-681), Fiberglass Plastic Carrier (B-883)
IIAR Abbreviation: (BLANK); Physical State: LIQ; Pressure Level: HIGH; Legend: Ammonia
24 x 4 in. (W x H)
3.5 in.
8 in.
Black, Orange, Red, Yellow
Ammonia (IIAR) Pipe Markers
Pipe and valve marking
609.6 x 101.6 mm (W x H)
88.9 mm
203.2 mm