Learn More
Brady™ Ammonia (IIAR) Pipe Markers, Legend: VAP/LOW/Ammonia, B-946 Self-Sticking Vinyl
For use in labeling ammonia piping and its contents. Brady Ammonia (IIAR) Pipe Markers help create a safer, more efficient work environment.
Specifications
Product Type Ammonia (IIAR) Pipe Marker
Color Black, Blue, Green, Yellow
Format Die-cut
Adhesive Type Acrylic, Permanent, Cold Temperature
Material Pressure Sensitive Vinyl (B-946)
View More Specs
Products 2
Catalog Number Mfr. No. Description Diameter (Metric) Height (Metric) Character Price Quantity & Availability  
Catalog Number Mfr. No. Description Diameter (Metric) Height (Metric) Character Price Quantity & Availability  
19-110-613
View Documents Promotion Details
Brady™
90469
Style I 76.2 to 127 mm 50.8 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
19-110-615
View Documents Promotion Details
Brady™
90471
Style IHV 203.2 mm 88.9 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Meets IIAR Standard and is recommended for use in labeling ammonia piping and its contents
  • Made of B-946 Self sticking pressure sensitive vinyl
  • Available in different style codes
Specifications
Specifications
Ammonia (IIAR) Pipe Marker
Die-cut
Pressure Sensitive Vinyl (B-946)
Pipe and Valve marking
Black, Blue, Green, Yellow
Acrylic, Permanent, Cold Temperature
IIAR Abbreviation: BLANK; Legend: Ammonia; Physical State: VAP; Pressure Level: LOW