Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonia Solution, FCC, 27-30%, Spectrum™

Click to view available options
Quantity:
500 mL
2.5 L
4 L
6 x 500 mL
20 L
4 x 2.5 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
Poly-Coated Glass Bottle
1336-21-6
H5NO
35.05
VHUUQVKOLVNVRT-UHFFFAOYSA-N
ammonium hydroxide
[NH4+].[OH-]
1336-21-6
H5NO
35.05
VHUUQVKOLVNVRT-UHFFFAOYSA-N
ammonium hydroxide
[NH4+].[OH-]

A1293, 1336-21-6, NH4OH

Spectrum™ Ammonia Solution, FCC is a solution of Ammonia in water. It has many uses, but in food production it is used primarily as a leavening agent. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Quantity 200 L
CAS Max % 1
UN Number UN2672
Lead (Pb) 0.5 mg/kg
Percent Purity 27 to 30%
Packaging Drum
Non_volatile Matter 0.0002