Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonia Solution, Strong, N.F. - F.C.C., J.T.Baker™

Click to view available options
Quantity:
2.5L
500mL
Packaging:
Glass Bottle

Chemical Identifiers

7664-41-7
H3N
17.031
QGZKDVFQNNGYKY-UHFFFAOYSA-N
222
CHEBI:16134
CAS 7664-41-7
Packaging Glass Bottle
Specific Gravity 1L = 0.90kg
Molecular Formula H3N
Quantity 500mL
InChI Key QGZKDVFQNNGYKY-UHFFFAOYSA-N
SMILES N
IUPAC Name azane
Molecular Weight (g/mol) 17.031
PubChem CID 222
ChEBI CHEBI:16134
Formula Weight 17.03
Chemical Name or Material Ammonia Solution, Strong
Grade FCC/NF
Show More Show Less