Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonia Solution, Strong, N.F. - F.C.C., J.T. Baker™

Catalog No. p-5391054
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle

Chemical Identifiers

CAS 7664-41-7
Molecular Formula H3N
Molecular Weight (g/mol) 17.03
MDL Number MFCD00011418
InChI Key QGZKDVFQNNGYKY-UHFFFAOYSA-N
Synonym ammonia, ammonia gas, spirit of hartshorn, nitro-sil, ammoniakgas, ammonia anhydrous, anhydrous ammonia, ammonia, anhydrous, ammoniak, am-fol
PubChem CID 222
ChEBI CHEBI:16134
SMILES N

Specifications

Quantity 500 mL
IUPAC Name ammonia
Formula Weight 17.03
Grade FCC/NF
Packaging Glass Bottle
Specific Gravity 1L = 0.90kg
Chemical Name or Material Ammonia Solution, Strong
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.