Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonia Standard, 1000ppm NH3, (822ppm N), Ricca Chemical

Click to view available options
Quantity:
120mL
500mL
1L
4L
Packaging:
Poly Bottle
12125-02-9,7732-18-5
ClH4N
53.489
NLXLAEXVIDQMFP-UHFFFAOYSA-N
25517
CHEBI:31206
azanium;chloride
[NH4+].[Cl-]
12125-02-9,7732-18-5
ClH4N
53.489
NLXLAEXVIDQMFP-UHFFFAOYSA-N
25517
CHEBI:31206
azanium;chloride
[NH4+].[Cl-]
CAS Min % 98.0,0.31
CAS Max % 100.0,0.32
Packaging Poly Bottle
Equivalence 1mL = 1mgNH3
Quantity 120mL
Chemical Name or Material Ammonia Standard, 1000ppm NH3