Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonia, T.S., BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
1L
Packaging:
Narrow Mouth Polyethylene Bottle

Chemical Identifiers

1336-21-6
H5NO
35.046
VHUUQVKOLVNVRT-UHFFFAOYSA-N
14923
CHEBI:18219
azanium;hydroxide
[NH4+].[OH-]

High quality chemicals for laboratory and specialized industrial use

Packaging Narrow Mouth Polyethylene Bottle
Specific Gravity 1L = 0.90kg
Quantity 1L
Chemical Name or Material Ammonia, T.S.
Grade BAKER ANALYZED™ Reagent