Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammoniacal Potassium Ferricyanide TS, (U.S.P. Test Solution), Spectrum™

Click to view available options
Quantity:
500 mL
1 L
Packaging:
Poly Bottle
1336-21-6,7732-18-5
H5NO
35.05
VHUUQVKOLVNVRT-UHFFFAOYSA-N
ammonium hydroxide
[NH4+].[OH-]
1336-21-6,7732-18-5
H5NO
35.05
VHUUQVKOLVNVRT-UHFFFAOYSA-N
ammonium hydroxide
[NH4+].[OH-]

A-296

Spectrum™ Ammoniacal Potassium Ferricyanide TS, (U.S.P. Test Solution) is a solution, sometimes designated with "TS or Test Solution", that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.
CAS Max % 0.1%,99.9%
UN Number UN2672
Packaging Poly Bottle
Quantity 500 mL