Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammoniated Mercury, Powder, USP, 98-100.5%, Spectrum™

missing translation for 'orderingAttributeHoverText'
Quantity:
125 g
500 g
2.5 kg
12 kg
Packaging:
Amber Glass Bottle
Poly Pail
10124-48-8
ClH2HgN
252.06
WRWRKDRWMURIBI-UHFFFAOYSA-M
mercury(2+) azanide chloride
[NH2-].[Cl-].[Hg++]
10124-48-8
ClH2HgN
252.06
WRWRKDRWMURIBI-UHFFFAOYSA-M
mercury(2+) azanide chloride
[NH2-].[Cl-].[Hg++]

AM168, 10124-48-8, Hg(NH2)Cl

Spectrum™ Ammoniated Mercury, Powder, USP is used to treat a variety of minor skin infections and other skin disorders. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Quantity 500 g
CAS Max % 1
UN Number UN1630
Percent Purity 98 to 100.5%