Ammonium Acetate - Acetic Acid Solution, TAPPI for Iron, Ricca Chemical

Manufacturer: Ricca Chemical CompanySupplier Diversity Partner 60.232

Packaging Poly natural
Quantity 1L

 View more versions of this product

Catalog No. 60-232

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7732-18-5,64-19-7,631-61-8
    CAS Min % 75.0,15.71,7.49
    CAS Max % 78.0,16.35,7.79
    Physical Form Liquid
    Chemical Name or Material Ammonium Acetate - Acetic Acid Solution

    Specifications

    Physical Form Liquid
    Color Clear colorless
    Quantity 1L
    Methods TAPPI for Iron
    CAS Max % 7.79,78.0,16.35
    Packaging Poly natural
    CAS Min % 75.0,15.71,7.49
    CAS 64-19-7,631-61-8,7732-18-5
    Shelf Life 24 Months
    Chemical Name or Material Ammonium Acetate - Acetic Acid Solution

    24 Months

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit