Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Acetate, Crystal, ACS, 97%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
90 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
631-61-8
631-61-8
Grade ACS
CAS Max % 1
Percent Purity 0.97
Chloride 5 ppm
Heavy Metals 5 ppm
Identification To Pass Test
Insoluble Matter 5e-05
Iron (Fe) 5 ppm
Nitrate 1e-05
Packaging Fiber Drum
Residue after Ignition 0.0001
Sulfate 1e-05
pH 6.7 to 7.3
Quantity 45 kg
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.