Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium Acetate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™
Click to view available options
Quantity:
500g
2kg
Packaging:
Poly Bottle
Specifications
Formula Weight | 77.08 |
Physical Form | Crystals |
Grade | BAKER ANALYZED™ ACS Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1L = 1.07kg |
Quantity | 2kg |
Chemical Name or Material | Ammonium Acetate |