Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Acetate, HPLC Grade, 97%, Spectrum™

Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly Bottle
Poly Pail
631-61-8
C2H7NO2
77.08
USFZMSVCRYTOJT-UHFFFAOYSA-N
ammonium acetate
[NH4+].CC([O-])=O
631-61-8
C2H7NO2
77.08
USFZMSVCRYTOJT-UHFFFAOYSA-N
ammonium acetate
[NH4+].CC([O-])=O

A2149, 631-61-8, C2H7NO2

Spectrum™ Ammonium Acetate, HPLC Grade is an inorganic salt used in the preparation of a buffered HPLC mobile phase. It is particularly useful when ionizable analytes are being investigated with reversed phase HPLC (RP-HPLC) to give better peak shape in the chromatogram. With a pKa value of 4.8, this material is a great selection when the required eluent pH is between 3.8 - 5.8 to achieve a particular separation. Spectrum Chemical manufactured HPLC grade salts meet the highest standards for quality and purity.
CAS Max % 1
Percent Purity 97%
Chloride 5 ppm
Heavy Metals (as Pb) 5 ppm
Insoluble Matter 5e-005
Nitrate 1e-005
Packaging Poly Pail
pH Range 6.7-7.3
Residue after Ignition 0.0001
Sulfate 1e-005
Quantity 1 kg
Show More Show Less