Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Acetate, Purified, Spectrum™

Click to view available options
Quantity:
25 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
631-61-8
631-61-8
Grade Purified
CAS Max % 1
Packaging Poly Bottle
Quantity 500 g

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.