Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Acetate TS, (U.S.P. Test Solution), Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Poly Bottle
631-61-8,7732-18-5
C2H7NO2
77.08
USFZMSVCRYTOJT-UHFFFAOYSA-N
ammonium acetate
[NH4+].CC([O-])=O
631-61-8,7732-18-5
C2H7NO2
77.08
USFZMSVCRYTOJT-UHFFFAOYSA-N
ammonium acetate
[NH4+].CC([O-])=O

A-299, 631-61-8

Spectrum™ Ammonium Acetate TS, (U.S.P. Test Solution) is a solution, sometimes designated with "TS or Test Solution", that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.
CAS Max % 0.1%,99.9%
Packaging Poly Bottle
Quantity 500 mL