Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Alum, Dodecahydrate, USP, 99-100.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
7784-26-1
AlH28NO20S2
453.31
WZUKKIPWIPZMAS-UHFFFAOYSA-K
aluminium(3+) ammonium dodecahydrate disulfate
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
7784-26-1
AlH28NO20S2
453.31
WZUKKIPWIPZMAS-UHFFFAOYSA-K
aluminium(3+) ammonium dodecahydrate disulfate
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

A1621, 7784-26-1, AlNH4(SO4)2•12H2O

Spectrum™ Ammonium Alum, Dodecahydrate, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Percent Purity 99 to 100.5%
Loss on Drying 45.0-48%
Packaging Poly Pail
Quantity 2.5 kg