Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ammonium benzoate, 99%

Manufacturer:  Alfa Aesar™ 3075336

Quantity 500g

 View more versions of this product

Catalog No. AA3075336


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1863-63-4
MFCD00013069
Benzoic acid ammonium salt
139.15
14,496
Ammonium benzoate

Specifications

1863-63-4
139.15
14,496
Weak
White to off-white
500g
Ammonium benzoate
Crystalline
MFCD00013069
Benzoic acid ammonium salt
Hygroscopic
192° to 198°C (dec.)
110° (230°F)
99%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P301+P312-P321-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Specific treatment (see ... on this label).Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : DG3378000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit