Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium bicarbonate, 98%, pure, ACROS Organics™

Click to view available options
Quantity:
25g
1kg
5kg
Packaging:
Glass bottle
Plastic Bottle
1066-33-7
CH5NO3
79.06
ATRRKUHOCOJYRX-UHFFFAOYSA-N
ammonium bicarbonate,ammonium hydrogencarbonate,ammonium hydrogen carbonate,carbonic acid, monoammonium salt,monoammonium carbonate,ammonium acid carbonate,ammonium hydrogencarbonat,ammoniumbicarbonate,acid ammonium carbonate,ccris 7327
14013
azanium;hydrogen carbonate
C(=O)(O)[O-].[NH4+]
1066-33-7
CH5NO3
79.06
ATRRKUHOCOJYRX-UHFFFAOYSA-N
ammonium bicarbonate,ammonium hydrogencarbonate,ammonium hydrogen carbonate,carbonic acid, monoammonium salt,monoammonium carbonate,ammonium acid carbonate,ammonium hydrogencarbonat,ammoniumbicarbonate,acid ammonium carbonate,ccris 7327
14013
azanium;hydrogen carbonate
C(=O)(O)[O-].[NH4+]
Assay Percent Range 98%
Linear Formula NH4HCO3
Merck Index 15,494
Solubility Information Solubility in water: 220g/L (20°C)
Formula Weight 79.06
Physical Form Crystalline Powder, Crystals, and/or Chunks
Percent Purity 98%
Grade Pure
Infrared Spectrum Authentic
Packaging Plastic Bottle
Color White
Melting Point 105°C
Quantity 5kg
Chemical Name or Material Ammonium bicarbonate
Show More Show Less

GHS H Statement
Harmful if swallowed.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Do not eat,drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.

GHS Signal Word: Warning

EINECSNumber : 213-911-5