Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Bicarbonate, Crystal, 99%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
1066-33-7
1066-33-7
Grade Reagent
CAS Max % 1
Percent Purity 0.99
Chloride 3e-05
Lead (Pb) 3 ppm
Packaging Poly Bottle
Sulfate 7e-05
Quantity 500 g

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.