Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Bicarbonate, FCC, 99-100.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
1066-33-7
C3H8N2O9Zn2
346.86
QOPSSYZLVIENBR-UHFFFAOYSA-J
dizinc(2+) diammonium tricarbonate
[NH4+].[NH4+].[Zn++].[Zn++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
1066-33-7
C3H8N2O9Zn2
346.86
QOPSSYZLVIENBR-UHFFFAOYSA-J
dizinc(2+) diammonium tricarbonate
[NH4+].[NH4+].[Zn++].[Zn++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

AM169, 1066-33-7, NH4HCO3

Spectrum™ Ammonium Bicarbonate, FCC is used as a raising agent in the food industry for flat baked goods, such as cookies and crackers. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Percent Purity 99 to 100.5%
Chloride 3e-005
Lead (Pb) 3 mg/kg
Non_volatile Matter 0.0055
Packaging Poly Pail
Sulfur (S) 7e-005
Quantity 2.5 kg