Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Bicarbonate, Powder, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle

Chemical Identifiers

1066-33-7
CH5NO3
79.055
ATRRKUHOCOJYRX-UHFFFAOYSA-N
14013
azanium;hydrogen carbonate
C(=O)(O)[O-].[NH4+]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 79.06
Physical Form Powder
Packaging Poly Bottle
Specific Gravity 1L = 1.59kg
Quantity 2.5kg
Chemical Name or Material Ammonium Bicarbonate
Grade BAKER ANALYZED™ Reagent