Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Bifluoride, Flake, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle
1341-49-7
NH4F·HF
1341-49-7
NH4F·HF

High quality chemicals for laboratory and specialized industrial use

Formula Weight 57.04
Physical Form Flakes
Grade BAKER ANALYZED™ Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.51kg
Quantity 2.5kg
Chemical Name or Material Ammonium Bifluoride