Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium Bifluoride, Purified, Reagents
Click to view available options
Quantity:
100 mL
500 g
3 kg
10 kg
25 kg
Specifications
Specifications
Boiling Point | 240°C |
Melting Point | 124.6°C |
Color | Colorless/White |
Physical Form | Solid |
Quantity | 25 kg |
Grade | Pure |
Packaging | Fiber Drum |
Specific Gravity | 1.5 |
Chemical Name or Material | Ammonium Bifluoride, Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction