Alfa Aesar™ Ammonium bis(oxalato)palladium(II) dihydrate, Premion™, 99.99% (metals basis)

Manufacturer: Alfa Aesar™ 4347506

Quantity 5g

 View more versions of this product

Catalog No. AA4347506

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula (NH4)2Pd(C2O4)2·2H2O
    Formula Weight 354.57 (318.54 Anhydrous)
    MDL Number MFCD02684521
    UN Number UN3288
    Chemical Name or Material Ammonium bis (oxalato)palladium (II) dihydrate
    Grade Premion™

    Specifications

    Formula Weight 354.57 (318.54 Anhydrous)
    Health Hazard 3 GHS P Statement
    P280-P301+P312-P312-P363-P322-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Wash contaminated clothing before reuse.
    Specific measures (see label).
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula (NH4)2Pd(C2O4)2·2H2O
    UN Number UN3288
    Quantity 5g
    Health Hazard 2 GHS H Statement
    H302-H312
    Harmful if swallowed.
    Harmful in contact with skin.
    Physical Form Crystalline
    MDL Number MFCD02684521
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
    Health Hazard 1 Warning
    Chemical Name or Material Ammonium bis (oxalato)palladium (II) dihydrate
    Assay 99.99%, (metals basis)
    Grade Premion™

    GHS H Statement
    H302-H312
    Harmful if swallowed.
    Harmful in contact with skin.

    GHS P Statement
    P280-P301+P312-P312-P363-P322-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Wash contaminated clothing before reuse.
    Specific measures (see label).
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit