Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Bisulfite, Reagent, 44-46%, Spectrum™

Click to view available options
Quantity:
200 L
Packaging:
Drum

A1145, 10192-30-0, H5NO3S

Spectrum™ Ammonium Bisulfite, 45 Percent Solution, Reagent
CAS 10192-30-0
CAS Max % 1
Molecular Formula H5NO3S
UN Number UN2693
InChI Key ZETCGWYACBNPIH-UHFFFAOYSA-N
SMILES N.OS(O)=O
IUPAC Name sulfurous acid amine
Molecular Weight (g/mol) 99.10
Percent Purity 44 to 46%
Packaging Drum
pH Range 4.5-6.5
Quantity 200 L
Show More Show Less