Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium bromide, 98%, extra pure, Acros Organics

Click to view available options
Quantity:
1kg
2.5kg
10kg
Packaging:
Plastic bottle
Plastic drum
12124-97-9
H4BrN
97.94
12124-97-9
H4BrN
97.94
Assay Percent Range 97.5% min. (Argentometry)
Linear Formula NH4Br
Merck Index 15,502
Solubility Information Solubility in water: 970g/L (25°C). Other solubilities: 1456g/L (100°C),freely soluble in alcohol and acetone,slightly soluble in ether,practically insoluble in ethyl acetate
Formula Weight 97.94
Percent Purity 98%
Grade Extra Pure
Insoluble Matter 0.01% max. (in water)
Packaging Plastic bottle
Solubility Solubility in water: 970g/L (25°C). Other solubilities: 1456g/L (100°C), freely soluble in alcohol and acetone, slightly soluble in ether, practically insoluble in ethyl acetate
Boiling Point 235.0°C
Melting Point 452.0°C
Quantity 2.5kg
Chemical Name or Material Ammonium bromide
Show More Show Less

GHS H Statement
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 235-183-8