Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium bromide, 99+%, for analysis, ACROS Organics™

Click to view available options
Quantity:
25g
250g
1kg
5kg
Packaging:
Glass bottle
Plastic bottle
12124-97-9
BrH4N
97.94
MFCD00011419
SWLVFNYSXGMGBS-UHFFFAOYSA-N
ammonium bromide
[NH4+].[Br-]
12124-97-9
BrH4N
97.94
MFCD00011419
SWLVFNYSXGMGBS-UHFFFAOYSA-N
ammonium bromide
[NH4+].[Br-]
Assay Percent Range 99% min. (on dry substance) (Argentometry)
Linear Formula NH4Br
Merck Index 15,502
Solubility Information Solubility in water: 970g/L (25°C). Other solubilities: 1456g/L (100°C),freely soluble in alcohol and acetone,slightly soluble in ether,practically insoluble in ethyl acetate
Barium (Ba) 20ppm max.
Formula Weight 97.94
Physical Form Crystalline Powder
Percent Purity 99+%
Grade Analysis
Bromate 0.001% max.
Chloride 0.2% max
Copper (Cu) 2ppm max.
Heavy Metals (as Pb) 10ppm max.
Iron (Fe) 1ppm max.
Lead (Pb) 2ppm max.
Loss on Drying 0.2% (105°C) max.
Nickel (Ni) 2ppm max.
Packaging Plastic bottle
Solubility Solubility in water: 970g/L (25°C). Other solubilities: 1456g/L (100°C), freely soluble in alcohol and acetone, slightly soluble in ether, practically insoluble in ethyl acetate
Sulfate 0.005% max
Sulfated Ash 0.05% max.
Boiling Point 235.0°C
Color White
Melting Point 452.0°C
pH 4.5 to 6.0 (5% soln.)
Quantity 250g
Chemical Name or Material Ammonium bromide
Show More Show Less

GHS H Statement
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 235-183-8