Learn More
Ammonium carbonate, ACS, NH{3} 30.0+%, Thermo Scientific Chemicals
Description
In baking powder, smelling salts, as an analytical lab reagent, manufacturing of rubber materials, casein glue, and fire extinguishing compoundsAmmonium carbonate is widely utilized as a leavening agent in lebkuchen, cookies and flat biscuits. It finds an important application as an emetic and an active ingredient
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- In baking powder, smelling salts, as an analytical lab reagent, manufacturing of rubber materials, casein glue, and fire extinguishing compounds
Specifications
Melting Point | 60° (sublimation) |
Physical Form | Powder |
Quantity | 500 g |
Assay Percent Range | NH3 30.0+% |
Molecular Formula | C2H11N3O5 |
Beilstein | 3627235 |
Merck Index | 14,508 |
Synonym | ammonium carbonate nf, ugam, ammonium bicarbonate-ammonium carbamate, ammonium carbonate chips nf, ammonium bicarbonate, mixt. with ammonium carbamate, ammonium carbamate, mixt. with ammonium bicarbonate, monoammonium carbonate mixture with ammonium carbamate, carbamic acid, monoammonium salt, mixt. with ammonium carbonate, diazanium hydrogen carbonate carbamate |
Solubility Information | Slowly soluble in water (1:4 portion); Decomposes in hot water. |
InChI Key | YNSUIDWLZCFLML-UHFFFAOYSA-N |
Show More |
Safety and Handling
Hazard Category | H302 |
Hazard Statement | GHS H Statement H302-H315-H319-H335 |
Precautionary Statement | P264b-P270-P301+P312-P330-P501c |
EINECSNumber | 208-058-0 |
RTECSNumber | BP1925000 |
TSCA | Yes |
storageNote1 | Hygroscopic |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.