Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium carbonate, extra pure, ACROS Organics™

Click to view available options
Quantity:
1kg
2.5kg
Packaging:
Plastic bottle
506-87-6
CH8N2O3
96.09
MFCD00010890
PRKQVKDSMLBJBJ-UHFFFAOYSA-N
ammonium carbonate,diammonium carbonate,carbonic acid, diammonium salt,ammonia sesquicarbonate,carbonic acid, ammonium salt,caswell no. 042,ammoniumcarbonat german,unii-pdp691cn28,carbonic acid, ammonium salt 1:2,ammoniumcarbonate
517111
diazanium;carbonate
C(=O)([O-])[O-].[NH4+].[NH4+]
506-87-6
CH8N2O3
96.09
MFCD00010890
PRKQVKDSMLBJBJ-UHFFFAOYSA-N
ammonium carbonate,diammonium carbonate,carbonic acid, diammonium salt,ammonia sesquicarbonate,carbonic acid, ammonium salt,caswell no. 042,ammoniumcarbonat german,unii-pdp691cn28,carbonic acid, ammonium salt 1:2,ammoniumcarbonate
517111
diazanium;carbonate
C(=O)([O-])[O-].[NH4+].[NH4+]
Linear Formula (NH4)2CO3
Merck Index 15, 505
Solubility Information Solubility in water: soluble (320g/L at 20°C)
Formula Weight 96.09
Physical Form Powder
Percent Purity ≥30.5% (NH3)
Grade Extra Pure
Chloride 30ppm max.
Heavy Metals (as Pb) 5ppm max.
Iron (Fe) 5ppm max.
Non_volatile Matter 100ppm max.
Packaging Plastic bottle
Solubility Solubility in water: soluble (320g/L at 20°C).
Sulfate 30ppm max.
Color White
Melting Point 58.0°C
Quantity 2.5kg
Chemical Name or Material Ammonium carbonate
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 208-058-