Ammonium Carbonate (Lumps/Certified ACS), Fisher Chemical

Packaging Poly Bottle
Quantity 3kg

 View more versions of this product

Catalog No. A656-3

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

Custom packing/Bulk ordering

You can now order this item in custom quantities.  Learn More

    Chemical Identifiers

    CAS 506-87-6
    Physical Form Solid
    Molecular Formula CH8N2O3
    Formula Weight 96.07
    MDL Number MFCD00085313
    Chemical Name or Material Ammonium Carbonate
    Assay ≥30%
    Grade Certified ACS Grade

    Specifications

    Formula Weight 96.07
    Melting Point 58°C
    Quantity 3kg
    Merck Index 15, 505
    Viscosity [NULL]
    Physical Form Solid
    Color White
    pH 9.4
    Decomposition Information 58°C
    Vapor Pressure 69 hPa at 20°C
    Health Hazard 3 Emergency Overview
    Harmful if swallowed. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.

    NFPA

    Health:2
    Flammability:1
    Instability:1
    Identification Pass Test
    Insoluble Matter 0.005% max.
    Sulfur Compounds 0.002% max.
    MDL Number MFCD00085313
    Health Hazard 2 WARNING!
    Chloride 5ppm max.
    Heavy Metals (as Pb) 5ppm max.
    Iron (Fe) 3ppm max.
    Non_volatile Matter 0.005% max.
    Packaging Poly Bottle
    Molecular Formula CH8N2O3
    CAS 506-87-6
    ChemAlert Storage Symbol Gray
    Additional Information Vapor Pressure: 69hPa at 20°C
    Chemical Name or Material Ammonium Carbonate
    Assay ≥30%
    Grade Certified ACS Grade
     

    WARNING!

    Emergency Overview
    Harmful if swallowed. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.

    NFPA

    Health:2
    Flammability:1
    Instability:1

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Product Certifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit