Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Carbonate (Lumps/HPLC), Fisher Chemical

Click to view available options
Quantity:
500g
Packaging:
Poly Bottle
506-87-6
CH8N2O3
96.086
MFCD00085313
PRKQVKDSMLBJBJ-UHFFFAOYSA-N
ammonium carbonate,diammonium carbonate,carbonic acid, diammonium salt,ammonia sesquicarbonate,carbonic acid, ammonium salt,caswell no. 042,ammoniumcarbonat german,unii-pdp691cn28,carbonic acid, ammonium salt 1:2,ammoniumcarbonate
517111
diazanium;carbonate
C(=O)([O-])[O-].[NH4+].[NH4+]
506-87-6
CH8N2O3
96.086
MFCD00085313
PRKQVKDSMLBJBJ-UHFFFAOYSA-N
ammonium carbonate,diammonium carbonate,carbonic acid, diammonium salt,ammonia sesquicarbonate,carbonic acid, ammonium salt,caswell no. 042,ammoniumcarbonat german,unii-pdp691cn28,carbonic acid, ammonium salt 1:2,ammoniumcarbonate
517111
diazanium;carbonate
C(=O)([O-])[O-].[NH4+].[NH4+]
Absorbance Pass Test
Additional Information Vapor Pressure: 69hPa at 20°C
Physical Form Solid
Percent Purity ≥30%
Grade Also meets ACS specifications
Chloride 5ppm max.
Heavy Metals 5ppm max.
Identification Pass Test
Insoluble Matter 0.005% max.
Iron (Fe) 3ppm max.
Non_volatile Matter 0.005% max.
Packaging Poly Bottle
Sulfur Compounds 0.002% max.
Vapor Pressure 69 hPa at 20°C
Decomposition Information 58°C
Color White
Melting Point 58°C
pH 9.4
Quantity 500g
Chemical Name or Material Ammonium Carbonate
Show More Show Less

WARNING!

Emergency Overview
Harmful if swallowed. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.

NFPA

Health:2
Flammability:1
Instability:1