Ammonium Carbonate (Lumps/NF), Fisher Chemical

Colorless, translucent lumps, 1“ (2.54cm) or smaller in size

Packaging Fiber Drum
Quantity 12kg

 View more versions of this product

Catalog No. A657-12

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

Custom packing/Bulk ordering

You can now order this item in custom quantities.  Learn More

    Chemical Identifiers

    CAS 506-87-6
    Physical Form Solid
    Assay Percent Range 30.0 to 34.0 %
    Molecular Formula CH8N2O3
    MDL Number MFCD00010890
    Chemical Name or Material Ammonium Carbonate
    Assay 30.0 to 34.0%
    Grade NF

    Specifications

    Melting Point 58°C
    Quantity 12kg
    Physical Form Solid
    Color White
    pH 9.4
    Decomposition Information 58°C
    Vapor Pressure 69 hPa at 20°C
    Health Hazard 3 Emergency Overview
    Harmful if swallowed. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.

    NFPA

    Health:2
    Flammability:1
    Instability:1
    Heavy Metals (as Pb) 0.001% max.
    Ignition Residue 0.05% max.
    Packaging Fiber Drum
    Identification Pass Test
    Assay Percent Range 30.0 to 34.0 %
    Molecular Formula CH8N2O3
    Sulfate 0.005% max.
    MDL Number MFCD00010890
    CAS 506-87-6
    ChemAlert Storage Symbol Gray
    Additional Information Vapor Pressure: 69hPa at 20°C
    Health Hazard 2 WARNING!
    Chloride 0.0035% max.
    Chemical Name or Material Ammonium Carbonate
    Assay 30.0 to 34.0%
    Grade NF
     

    WARNING!

    Emergency Overview
    Harmful if swallowed. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.

    NFPA

    Health:2
    Flammability:1
    Instability:1

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Product Certifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit