Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

Ammonium Carbonate (Lumps/NF), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.

Colorless, translucent lumps, 1“ (2.54cm) or smaller in size

Packaging Poly Bottle
Quantity 500g

 View more versions of this product

Catalog No. A657-500



 
Add to cart

Custom packing/Bulk ordering

You can now order this item in custom quantities. Learn More

Specifications Description & Specifications

Chemical Identifiers

506-87-6
30.0 to 34.0 %
MFCD00010890
30.0 to 34.0%
Solid
CH8N2O3
Ammonium Carbonate
NF

Specifications

506-87-6
Solid
CH8N2O3
0.0035% max.
Pass Test
Poly Bottle
69 hPa at 20°C
White
9.4
Ammonium Carbonate
NF
Vapor Pressure: 69hPa at 20°C
30.0 to 34.0 %
MFCD00010890
0.001% max.
0.05% max.
0.005% max.
58°C
58°C
500g
30.0 to 34.0%

Safety and Handling

SDS & Safety Documents

WARNING!

Emergency Overview
Harmful if swallowed. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.

NFPA

Health:2
Flammability:1
Instability:1

Alternative Products
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit