Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Carbonate, Powder, FCC, 30-34%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
8000-73-5
C2H11N3O5
157.13
YNSUIDWLZCFLML-UHFFFAOYSA-N
diammonium carbamate hydrogen carbonate
[NH4+].[NH4+].NC([O-])=O.OC([O-])=O
8000-73-5
C2H11N3O5
157.13
YNSUIDWLZCFLML-UHFFFAOYSA-N
diammonium carbamate hydrogen carbonate
[NH4+].[NH4+].NC([O-])=O.OC([O-])=O

AM173, 8000-73-5

Spectrum™ Ammonium Carbonate, Powder, FCC is a white, ammoniacal, solid salt that is used as a pH adjuster and as a cooking leavening agent. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Percent Purity 30 to 34%
Chloride 3e-005
Lead (Pb) 3 mg/kg
Non_volatile Matter 0.0005
Packaging Poly Pail
Sulfur (S) 5e-005
Quantity 12 kg