Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Carbonate, Powder, NF, 30-34%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
8000-73-5
C2H11N3O5
157.13
YNSUIDWLZCFLML-UHFFFAOYSA-N
diammonium carbamate hydrogen carbonate
[NH4+].[NH4+].NC([O-])=O.OC([O-])=O
8000-73-5
C2H11N3O5
157.13
YNSUIDWLZCFLML-UHFFFAOYSA-N
diammonium carbamate hydrogen carbonate
[NH4+].[NH4+].NC([O-])=O.OC([O-])=O

A1630, 8000-73-5

Spectrum™ Ammonium Carbonate, Powder, NF is an excipient used as an alkalinizing agent in liquid preparations for product stability. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Percent Purity 30 to 34%
Chloride 0.0035%
Ignition Residue 0.1%
Packaging Poly Pail
Sulfate 0.005%
Quantity 12 kg