Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell™ Ammonium Cerium(Iv) Nitrate, ≥98.0% (T), purum p.a., Honeywell™
Click to view available options
Quantity:
100g
500g
Packaging:
Poly Bottle
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications
Linear Formula | Ce(NH4)2(NO3)6 |
UN Number | UN3085 |
Formula Weight | 548.22g/mol |
Percent Purity | ≥98.0% (T) |
Grade | purum p.a. |
Packaging | Poly Bottle |
Quantity | 100g |
Chemical Name or Material | Ammonium Cerium(Iv) Nitrate |
Safety and Handling
P210-P220-P260-P280-P305 + P351 + P338-P370 + P378
H272-H290-H302-H314-H317-H410
DOTInformation : Class 5.1 - PG 2 -EHS - acidic - Oxidizing solid, corrosive, n.o.s., HI: all
EINECSNumber : 240-827-6