Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium Cerium(Iv) Nitrate, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 500 g |
Linear Formula | Ce(NH4)2(NO3)6 |
UN Number | 3085 |
Formula Weight | 548.22g/mol |
Percent Purity | ≥98.5% (RT) |
Grade | ACS/puriss. p.a. |
Packaging | Plastic Bottle |
Chemical Name or Material | Ammonium Cerium(Iv) Nitrate |
Safety and Handling
P210-P220-P260-P280-P305 + P351 + P338-P370 + P378
H272-H290-H302-H314-H317-H410
EINECSNumber : 240-827-6