Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium cerium(IV) sulfate dihydrate, 99+%, extra pure, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
Plastic bottle
10378-47-9
H16CeN4O16S4·2H2O
632.54
MFCD00149417
Ceric ammonium sulfate
10378-47-9
H16CeN4O16S4·2H2O
632.54
MFCD00149417
Ceric ammonium sulfate
Assay Percent Range 99+%
Linear Formula (NH4)4Ce(SO4)4·2H2O
Fieser 08,80
Solubility Information Solubility in water: hydrolysis. Other solubilities: insoluble in ethanol
Formula Weight 632.54
Physical Form Crystalline Powder
Percent Purity 99+%
Grade Extra Pure
Chloride 100ppm max
Chlorine (Cl) 100ppm max.
Insoluble Matter 0.02% max. (in dilute H2SO4)
Iron (Fe) 50ppm max.
Packaging Plastic bottle
Phosphate 100ppm max.
Solubility Solubility in water: hydrolysis. Other solubilities: insoluble in ethanol
Color Orange
Melting Point 130.0°C
Quantity 100g
Chemical Name or Material Ammonium cerium(IV) sulfate dihydrate, Extra Pure
Show More Show Less

GHS Signal Word: Warning