Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Chloride, 10% (w/v) Solution, Spectrum™

Click to view available options
Quantity:
500 mL
1 L
Packaging:
Poly Bottle
12125-02-9
12125-02-9
Grade Chemical Solution
Packaging Poly Bottle
Quantity 1 L