Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium chloride, 99.5%, extra pure, ACROS Organics™

Click to view available options
Quantity:
500g
1kg
5kg
25kg
Packaging:
Cardboard Drum
Plastic bottle
Plastic drum
12125-02-9
H4ClN
53.49
MFCD00011420
NLXLAEXVIDQMFP-UHFFFAOYSA-N
ammonium chloride,salmiac,sal ammoniac,ammonium muriate,ammoniumchlorid,ammoneric,darammon,amchlor,ammonium chloride nh4 cl,ammoniumklorid
25517
CHEBI:31206
azanium;chloride
[NH4+].[Cl-]
12125-02-9
H4ClN
53.49
MFCD00011420
NLXLAEXVIDQMFP-UHFFFAOYSA-N
ammonium chloride,salmiac,sal ammoniac,ammonium muriate,ammoniumchlorid,ammoneric,darammon,amchlor,ammonium chloride nh4 cl,ammoniumklorid
25517
CHEBI:31206
azanium;chloride
[NH4+].[Cl-]
Assay Percent Range 99.5% min. (Argentometry)
Linear Formula NH3·HCl
Fieser 15,13
Merck Index 15, 506
Solubility Information Solubility in water: 370g/L (20°C). Other solubilities: soluble in glycerol,slightly soluble in alcohol,practically insoluble in acetone,ether and,ethyl acetate
Formula Weight 53.49
Physical Form Powder
Percent Purity 99.5%
Grade Extra Pure
Density 1.5200g/mL
Heavy Metals (as Pb) 5ppm max.
Insoluble Matter 0.005% max
Iron (Fe) 2ppm max.
Packaging Plastic bottle
Phosphate 2ppm max.
Solubility Solubility in water: 370g/L (20°C). Other solubilities: soluble in glycerol, slightly soluble in alcohol, practically insoluble in acetone, ether and, ethyl acetate
Sulfate 20ppm max.
Sulfated Ash 0.01% max.
Sublimation Point 0160.0°C
Color White
Melting Point 340.0°C
pH 4.5 to 5.5 (5% soln. at 20°C)
Quantity 1kg
Chemical Name or Material Ammonium chloride
Show More Show Less

GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Do not eat,d

GHS Signal Word: Warning

EINECSNumber : 235-186-4