Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium chloride, 99.5%, extra pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
1 kg
5 kg
25 kg
Packaging:
Fiber Drum
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 340.0°C |
Color | White |
pH | 4.5 to 5.5 (5% soln. at 20°C) |
Physical Form | Powder |
Quantity | 25 kg |
Assay Percent Range | 99.5% min. (Argentometry) |
Linear Formula | NH3·HCl |
Fieser | 15,13 |
Merck Index | 15, 506 |
Solubility Information | Solubility in water: 370g/L (20°C). Other solubilities: soluble in glycerol,slightly soluble in alcohol,practically insoluble in acetone,ether and,ethyl acetate |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement Causes serious eye irritation. Harmful if swallowed. |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Do not eat,d |
EINECSNumber | 235-186-4 |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction