Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Chloride, +99%, Pure, ACROS Organics™

Click to view available options
Quantity:
25g
1kg
5kg
25kg
Packaging:
Fibre drum
Plastic Bottle
Plastic drum
12125-02-9
H4ClN
53.49
MFCD00011420
NLXLAEXVIDQMFP-UHFFFAOYSA-N
ammonium chloride,salmiac,sal ammoniac,ammonium muriate,ammoniumchlorid,ammoneric,darammon,amchlor,ammonium chloride nh4 cl,ammoniumklorid
25517
CHEBI:31206
azanium;chloride
[NH4+].[Cl-]
12125-02-9
H4ClN
53.49
MFCD00011420
NLXLAEXVIDQMFP-UHFFFAOYSA-N
ammonium chloride,salmiac,sal ammoniac,ammonium muriate,ammoniumchlorid,ammoneric,darammon,amchlor,ammonium chloride nh4 cl,ammoniumklorid
25517
CHEBI:31206
azanium;chloride
[NH4+].[Cl-]
Assay Percent Range 99+%
Linear Formula NH3·HCl
Fieser 15,13
Merck Index 15, 506
Solubility Information Solubility in water: 370g/L (20°C). Other solubilities: soluble in glycerol,slightly soluble in alcohol,practically insoluble in acetone,ether and,ethyl acetate
Formula Weight 53.49
Physical Form Crystalline Powder and Chunks
Percent Purity ≥99%
Grade Pure
Density 1.5200g/mL
Insoluble Matter 0.02% max.
Iron (Fe) 10ppm max.
Packaging Plastic Bottle
Solubility Solubility in water: 370g/L (20°C). Other solubilities: soluble in glycerol, slightly soluble in alcohol, practically insoluble in acetone, ether and, ethyl acetate
Sulfated Ash 0.3% max.
Water 0.1% max. (K.F.)
Sublimation Point 0160.0°C
Color White
Melting Point 340.0°C
Quantity 25g
Chemical Name or Material Ammonium chloride
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face, hands and any exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning