Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Chloride/Ammonium Hydroxide Buffer (Copper Buffer), Reagents

Catalog No. 01335960
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L

Ammonium Chloride/Ammonium Hydroxide Buffer (Copper Buffer), Reagent, use for EDTA titrations. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Ammonium Chloride/Ammonium Hydroxide Buffer (Copper Buffer)
CAS 7732-18-5, 1336-21-6, 1336-21-6
Color Colorless
Concentration 1 M
For Use With (Application) Titration
pH 11.5 to 12.5
Physical Form Liquid
Quantity 20 L
Solution Type Buffer
Packaging Cubitainer
Specific Gravity 1.0185
Sterility Non-sterile
Grade Reagent
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.